Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-C 0801,B2M Monomer Protein HLM-H82Es is expressed from human 293 cells HEK293. It contains AA Cys 25 - Ile 30 8 HLA-C 0801, Ile 21 - Met 119 B2M Accession QOC77848.1 HLA-C 0801, P61769 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82ES-25UG
Supplier:  ACRO Biosystems
Biotinylated Human HLA-C 0801,B2M Monomer Protein HLM-H82Es is expressed from human 293 cells HEK293. It contains AA Cys 25 - Ile 30 8 HLA-C 0801, Ile 21 - Met 119 B2M Accession QOC77848.1 HLA-C 0801, P61769 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82ES-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together