Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-B 5701,B2M Monomer Protein HLM-H82Ef is expressed from human 293 cells HEK293. It contains AA Gly 25 - Val 30 9 HLA-B 0801, Ile 21-Met 119 B2M Accession CAD7343887.1 HLA-B 5701, P61769-1 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82EF-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human HLA-B 5701,B2M Monomer Protein HLM-H82Ef is expressed from human 293 cells HEK293. It contains AA Gly 25 - Val 30 9 HLA-B 0801, Ile 21-Met 119 B2M Accession CAD7343887.1 HLA-B 5701, P61769-1 B2M. 1 * 25 µG
Catalog # ACRBHLM-H82EF-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together