Order Entry
Poland
ContactUsLinkComponent
Unconjugated Human HLA-A 0201,B2M Monomer Protein HLM-H52H5 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Ile 30 8 HLA-A 0201, Ile 21 - Met 119 B2M Accession AAA59606.1 HLA-A 0201, P61769-1 B2M. 1 * 25 µG
  ACRBHLM-H52H5-25UG
Unconjugated Human HLA-A 0201,B2M Monomer Protein HLM-H52H5 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Ile 30 8 HLA-A 0201, Ile 21 - Met 119 B2M Accession AAA59606.1 HLA-A 0201, P61769-1 B2M. 1 * 25 µG
  ACRBHLM-H52H5-25UG
Frequently Bought Together