Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-DRA1 0101,HLA-DRB1 0401 Monomer Protein HL1-H82W8 is expressed from human 293 cells HEK293. It contains AA Il e 26 - Glu 216 HLA-DRA1 0101, Gly 30 - Lys 227 HLA-DRB1 0401 Accession CAI2388006.1 HLA-DRA1 0101, CAI9239961.1 HLA- 1 * 25 µG
  ACRBHL1-H82W8-25UG
Biotinylated Human HLA-DRA1 0101,HLA-DRB1 0401 Monomer Protein HL1-H82W8 is expressed from human 293 cells HEK293. It contains AA Il e 26 - Glu 216 HLA-DRA1 0101, Gly 30 - Lys 227 HLA-DRB1 0401 Accession CAI2388006.1 HLA-DRA1 0101, CAI9239961.1 HLA- 1 * 25 µG
  ACRBHL1-H82W8-25UG
Frequently Bought Together