Order Entry
Poland
ContactUsLinkComponent
Biotinylated Human HLA-A 3303,B2M,EGF R HVKITDFGR Complex Protein HLR-H82E3 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Ile 308 HLA-A 3303, Ile 21 - Met 119 B2M, HVKITDFGR peptide Accession AAA79865.1 HLA-A 3303, P61769-1 B2M, 1 * 25 µG
Catalog # ACRBHLR-H82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human HLA-A 3303,B2M,EGF R HVKITDFGR Complex Protein HLR-H82E3 is expressed from human 293 cells HEK293. It contains AA Gly 25 - Ile 308 HLA-A 3303, Ile 21 - Met 119 B2M, HVKITDFGR peptide Accession AAA79865.1 HLA-A 3303, P61769-1 B2M, 1 * 25 µG
Catalog # ACRBHLR-H82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together