Pterostilbene 98%
Glass bottle, 5 g
Catalog # 471380050.
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
- CAS Number:537-42-8
- Formula:C₁₆H₁₆O₃
- MDL Number:MFCD00238710
- Molecular Weight:256.2964 g/mol
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
Specifications
Specification Test Results
- Appearance:white to beige crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:90°C to 98°C
- Water:≤3 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



