Order Entry
Poland
Orders LinkContactUsLinkComponent
 

3,3'-Dihydroxybenzidine 98%

Glass bottle, 5 g
Catalog # 474450050.
Health
3,3'-Dihydroxybenzidine 98%
Catalog # 474450050.
CAS Number:  2373-98-0
3,3'-Dihydroxybenzidine 98%

Glass bottle, 5 g

CAS Number:  2373-98-0
Price
512 zł
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
  • CAS Number:2373-98-0
  • Formula:C₁₂H₁₂N₂O₂
  • MDL Number:MFCD00039149
  • Molecular Weight:216.24 g/mol
  • Storage Temperature:Ambient
  • EINECS:220-190-0
Specifications

Specification Test Results

  • Appearance:
    white to brown powder or crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications