SARS-CoV-2 (COVID-19) Nucleocapsid Monoclonal Antibody [N008] 1 * 0,1 mg
Catalog # PRSI10-542
Supplier: ProSci Inc.
SARS-CoV-2 (COVID-19) Nucleocapsid Monoclonal Antibody [N008] 1 * 0,1 mg
Catalog # PRSI10-542
Supplier: ProSci Inc.
Frequently Bought Together
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Cell culture storage bottles, polystyrene, Corning®
Supplier: Corning
Cryoboxes, B47/B67/B77/B97-series
Supplier: GLW
Spare Closures for VWR® Media Bottles
Supplier: VWR Collection
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: STERILIN
Blotting transfer papers
Supplier: Ahlstrom-Munksjö
VWR®, Inoculating Loops and Needles, Disposable
Supplier: VWR Collection