SARS-CoV-2 (COVID-19) pp1ab Antibody (NSP3-IN2) 1 * 0,1 mg
PRSI10-506
Frequently Bought Together
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR® Standard Line Transfer Pipettes
: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Transfer Pipettes, Graduated
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection