High Lunatic Plates, OD 275 (Box of 40). Box of 40 High Lunatic Plates, 0.03-275 OD. 96 samples per plate. For use with Lunatic (Lunatic & Stunner Client v8.3 or higher) 1 * 40 items
Catalog # MSPP701-2101
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together