RSV RECOMBINANT MONOCLONAL ANTIBODY (RSHZ19) (RABBIT IGGΚ) 1 * 200 µG
ENZOENZABS6560200
RSV RECOMBINANT MONOCLONAL ANTIBODY (RSHZ19) (RABBIT IGGΚ) 1 * 200 µG
ENZOENZABS6560200
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR® Blender Bags
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection