Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IL-6 RECEPTOR RECOMBINANT MONOCLONAL ANTIBODY (RHPM-1) (RABBIT IGGΚ) 1 * 200 µG
Catalog # ENZOENZABS5500200
Supplier:
Enzo Life Sciences
Expand image
IL-6 RECEPTOR RECOMBINANT MONOCLONAL ANTIBODY (RHPM-1) (RABBIT IGGΚ) 1 * 200 µG
Catalog # ENZOENZABS5500200
Supplier:
Enzo Life Sciences
Frequently Bought Together
VWR® Blender Bags
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection