Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
NINTEDANIB (FREE BASE) 1 * 100 mg
Catalog # ENZOENZCHM2300100
Supplier:
Enzo Life Sciences
Expand image
NINTEDANIB (FREE BASE) 1 * 100 mg
Catalog # ENZOENZCHM2300100
Supplier:
Enzo Life Sciences
Frequently Bought Together
Accessories for photometric test kits, NANOCONTROL
Supplier: MACHEREY-NAGEL
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Supplier: Honeywell Chemicals
FIXATIVE CAL-RITE 1GAL 1 * 1 items
Supplier: SHANDON
EOSIN RAL 555 2.5L 1 * 2,5 L
Supplier: RAL