Przejdź do Nagłówek
Przejdź do Główna treść
Przejdź do Stopka
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Wprowadź zamówienie
Status zamówienia
Twoje konto
Koszyk
Poland
Wprowadź zamówienie
Status zamówienia
Twoje konto
Koszyk
ContactUsLinkComponent
PROTEASOME 19S ATPASE SUBUNIT RPT6 MONOCLONAL ANTIBODY (P45-110) 1 * 25 µl
Catalog # ENZOBMLPW92650025
Supplier:
ENZO LIFE SCIENCES
Expand image
PROTEASOME 19S ATPASE SUBUNIT RPT6 MONOCLONAL ANTIBODY (P45-110) 1 * 25 µl
Catalog # ENZOBMLPW92650025
Supplier:
ENZO LIFE SCIENCES
Frequently Bought Together
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection