Order Entry
Poland
Orders LinkContactUsLinkComponent
Midostaurin ≥98% (by TLC)

Midostaurin ≥98% (by TLC)

5 mg
Catalog # ENZOBMLEI3700005
Supplier:   Enzo Life Sciences
Midostaurin ≥98% (by TLC)
Catalog # ENZOBMLEI3700005
Supplier:  Enzo Life Sciences
CAS Number:  120685-11-2
Midostaurin ≥98% (by TLC)
Midostaurin ≥98% (by TLC)

5 mg

CAS Number:  120685-11-2
Price
2 770 zł
Log in for availability
1 * 5 mg
Product Details & Documents

Inhibitor of a variety of serine/threonine and tyrosine kinases, like protein kinase C (PKC), cyclic AMP-dependent protein kinase (PKA), S6 kinase, Akt (protein kinase B; PKB), epidermal growth factor receptor (EGFR) tyrosine kinase activity and others including KDR, VEGFR, PDGFR, c-kit and other receptor tyrosine kinases. Potently inhibits FLT-3 kinase including mutant forms found in acute myeloid leukemia in vitro and in vivo. Apoptosis inducer. Showed broad antiproliferative activity against various tumor cell lines. Selectively inhibits T lymphocyte production of TNF-α. Upregulates endothelial nitric oxide synthase (eNOS; NOS III). Abrogates tumor angiogenesis in vivo.

  • Flt inhibitor

White to off-white powder.

Soluble in DMSO (15mg/ml), MDC (10mg/ml), methanol (5mg/ml) or 100% ethanol (2.5mg/ml).

Synonyms:  PKC412, N-((5S,6R,7R,9R)-6-Methoxy-5-methyl-14-oxo-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-7-yl)-N-methylbenzamide
  • CAS Number:120685-11-2
  • Formula:C₃₅H₃₀N₄O₄
  • MDL Number:MFCD12828879
  • Molecular Weight:570.64 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    5 mg

Specifications