Odczynniki do ekstrakcji
Extraction reagent 1, 5X, 10 mL
Catalog # ENZOADI801526
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Concentrated lysis buffer for preparation of cell and tissue extracts.
TRIS buffer containing detergents.
Specifications
Specifications
- Ilość:10 mL
- Opis:Extraction reagent 1, 5X