Order Entry
Poland
ContactUsLinkComponent
BUFFER PH3 EXP FOR VWR HITACHI AAA - Buffer without Ethanol - Ethanol has to been added before usage 1 * 1 L
Catalog # HPLCB17001EXP
BUFFER PH3 EXP FOR VWR HITACHI AAA - Buffer without Ethanol - Ethanol has to been added before usage 1 * 1 L
Catalog # HPLCB17001EXP
Frequently Bought Together