Nunc™ EasYFlask™ Flasks with Nunclon™ Supra Surface
Product Details & Documents
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
- Consistent lot-to-lot performance
- Sterile with SAL 10⁻²
- Non-cytotoxic
- Non-pyrogenic
- Biocompatible per USP Class VI for raw materials
- Shelf life: Five years
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Specifications
- Closure type:Solid cap
- Packed:200 flasks per case
- Description:Nunc EasYFlask, 25 cm²
- Seq:02
- Working volume:7 ml
- Cat. No.:734-3648
- Environmental certification:IRECS
- Environmentally Preferable:Low Manufacturing Impact



