FMOC-ASP(OTBU)-CYS(PSI(DMP,H)PRO)-OH 1 * 250 mg
Catalog # APOSOR36493-250MG
Supplier: Apollo Scientific
FMOC-ASP(OTBU)-CYS(PSI(DMP,H)PRO)-OH 1 * 250 mg
Catalog # APOSOR36493-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Methanol ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Chloroform stabilised, pure analytical reagent
Supplier: CHEMPUR
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Trifluoroacetic Acid for UV 1 * 100 mL
Supplier: PanReac AppliChem
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
Methanol, pure
Supplier: CHEMPUR
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Dimethyl sulphoxide ≥99.9% for HPLC
Supplier: Thermo Fisher Scientific