Skip to Header
Skip to Main Content
Skip to Footer
Zaawansowany
Zaawansowany
SDS
Certyfikaty
Dane producenta
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1 component: 1,2,3-Trichloropropane [CAS:96-18-4] 2000mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0082201.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
1 component: 1,2,3-Trichloropropane [CAS:96-18-4] 2000mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0082201.5
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Total Iron Test Strips, Dosatest, VWR®
Supplier: VWR Chemicals
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
VISOCOLOR® Colorimetric Test, Nitrate
Supplier: MACHEREY-NAGEL
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
VWR® Powder Packs for Determination of Iron, Reagents for Photometers
Supplier: VWR Chemicals
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Ammonium acetate, Supelco®
Supplier: Merck
Colorimetric Rapid Tests, Aluminium, VISOCOLOR®
Supplier: MACHEREY-NAGEL