10-Bromo-5H-dibenz[b,f]azepine-5-carboxamide certified reference material, TraceCERT(R) 1 * 100 mg
Catalog # PHR9066-100MG
Supplier: Merck
10-Bromo-5H-dibenz[b,f]azepine-5-carboxamide certified reference material, TraceCERT(R) 1 * 100 mg
Catalog # PHR9066-100MG
Supplier: Merck
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Osmolality standards
Supplier: VWR Chemicals
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
Supelco® Organic Reference Standard, Dihexyl phthalate
Supplier: Merck
VWR®, Cone and Plate viscosity standards
Supplier: VWR Chemicals
Disulfiram, Reference Standard
Supplier: USP
Ethanolamine, Supelco®
Supplier: Merck
pH buffer solutions, colour coded
Supplier: Hach
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Scandium standard solution, 1000 mg/L Sc in 5% nitric acid (from Sc₂O₃), standard for AAS, Specpure®
Supplier: Thermo Fisher Scientific
Vardenafil Related Compounds, Reference Standard
Supplier: USP
ARCTIIN, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck
MULTI ELEMENT STD 75 BLANK OIL S-21:30 1 * 200 g
Supplier: LGC Standards PROMOCHEM