Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid certified reference material, TraceCERT(R) 1 * 100 mg
Catalog # PHR9010-100MG
Supplier:
Merck
Expand image
1-Ethyl-6-fluoro-7-(4-formylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid certified reference material, TraceCERT(R) 1 * 100 mg
Catalog # PHR9010-100MG
Supplier:
Merck
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Chloride standard solution, 1,000 mg/L Cl- in Water (from NaCl) Certipur®, Supelco®
Supplier: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Mercury standard solution, 1000 mg/L Hg in 2 M nitric acid (from Hg(NO₃)₂) Certipur® standard for AAS, Supelco®
Supplier: Merck
Redox buffer solutions
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffer solutions, colour coded
Supplier: Hach