Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Aminolevulinic Acid Related Compound A pharmaceutical secondary standard, certified reference material 1 * 50 mg
Catalog # PHR2995-50MG
Supplier:
Merck
Expand image
Aminolevulinic Acid Related Compound A pharmaceutical secondary standard, certified reference material 1 * 50 mg
Catalog # PHR2995-50MG
Supplier:
Merck
Frequently Bought Together
Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution for filling electrodes, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Astaxanthin ?97% (HPLC), from Haematococcus pluvailis 1 * 1 items
Supplier: Merck
VWR®, Cone and Plate viscosity standards
Supplier: VWR Chemicals
Buffer solutions
Supplier: REAGECON
MULTI-ELEMENT CALIBRATION STANDARD 2? 1 * 1 items
Supplier: PerkinElmer
Ganciclovir, Reference Standards
Supplier: USP
2-Methyl-1-propanol, Reference Standard
Supplier: USP
Residual Solvent - Acetonitrile Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Supplier: Merck