Residual Solvent - Methanol Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Catalog # PHR1170-3X1.2M
Supplier: Merck
Residual Solvent - Methanol Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Catalog # PHR1170-3X1.2M
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, 2,2,2-Trifluoroethanol
Supplier: Merck
Hard Drinking Water UK Low Level Metals
Supplier: LGC Standards PROMOCHEM
Supelco® Organic Reference Standard, α-Tocotrienol
Supplier: Merck
Vardenafil Related Compounds, Reference Standard
Supplier: USP
Conductivity standards, KCl, referenced at 25 °C
Supplier: Thermo Fisher Scientific
PHENYTOIN INTERMEDIATE COMPOUND M01 1 * 100 mg
Supplier: Merck
Phthalic anhydride 160 g/l in pyridine, BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Rotigotine Related Compounds, Reference Standard
Supplier: USP
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
pH buffer solutions, colour coded
Supplier: Hach