Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pli able in extreme temperatures. All labels can be customized or left blank. 1 * 20 items
Catalog # VELTC2WR-NB-02
Supplier: VELTEK ASSOCIATES
Frequently Bought Together