GLYCOPYRRONIUM BROMIDE - IMPURITY B 1 * 25 mg
MSPPPA0713020
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo
UV/Visible standards, Supelco®
: Merck
pH buffers, ready to use
: WTW
VWR®, Conductivity standard solutions
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo