0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
The E.Z.N.A.® Blood DNA Kit provides rapid total DNA isolation from 1-250 µL of fresh and frozen anticoagulated whole blood. 1 * 600 Tests
Catalog # OMEGD3392-03
Supplier:
OMEGA BIO-TEK
Expand image
The E.Z.N.A.® Blood DNA Kit provides rapid total DNA isolation from 1-250 µL of fresh and frozen anticoagulated whole blood. 1 * 600 Tests
Catalog # OMEGD3392-03
Supplier:
OMEGA BIO-TEK
Frequently Bought Together
VWR®, Accessories for VWR Bead Mill Homogenisers
Supplier: VWR Collection
Flow Cell Priming Kit
Supplier: Oxford Nanopore Technologies
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies
Control Expansion
Supplier: Oxford Nanopore Technologies
Rapid sequencing kits
Supplier: Oxford Nanopore Technologies
cDNA-PCR Sequencing Kits
Supplier: Oxford Nanopore Technologies
16S Barcoding Kits
Supplier: Oxford Nanopore Technologies
Sequencing auxiliary vials
Supplier: Oxford Nanopore Technologies