Order Entry
Poland
ContactUsLinkComponent
SHODEX GPC HK-402, ULTRA-RAPID SIZE EXCLUSION COLUMN, 4,6x150mm, 3µm, 300Å, THF 1 * 1 items
Catalog # SHOWF6025020
Supplier:  Resonac
SHODEX GPC HK-402, ULTRA-RAPID SIZE EXCLUSION COLUMN, 4,6x150mm, 3µm, 300Å, THF 1 * 1 items
Catalog # SHOWF6025020
Supplier:  Resonac
Frequently Bought Together