Order Entry
Poland
ContactUsLinkComponent
Cynomolgus IL-2RB&IL-2RA&IL-2RG, His Tag&Twin-Strep Tag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 30.1 kDa (IL-2RB) and 55.9 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a his tag at the C-terminu 1 * 1 mg
Catalog # ACRBILG-C52W3-1MG
Supplier:  ACRO Biosystems
Cynomolgus IL-2RB&IL-2RA&IL-2RG, His Tag&Twin-Strep Tag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 30.1 kDa (IL-2RB) and 55.9 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a his tag at the C-terminu 1 * 1 mg
Catalog # ACRBILG-C52W3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together