[10-(NAPHTHALENE-1-YL)-9-ANTHRACENEYL]BO 1 * 5 g
Catalog # APOSOR315369-5G
Supplier: Apollo Scientific
[10-(NAPHTHALENE-1-YL)-9-ANTHRACENEYL]BO 1 * 5 g
Catalog # APOSOR315369-5G
Supplier: Apollo Scientific
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
2-ETHOXYETHANOL, REAGENTPLUS, 99%. 1 * 1 Each
Supplier: Merck
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
ACETON CZDA-BASIC 99,5% OP. 1 DM3 1 * 1 L
Supplier: POCH AVANTOR BRAND
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
n-Hexane for HPLC
Supplier: POCH AVANTOR BRAND
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Isoamyl alcohol mixture of isomers ≥99%
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific