PROPARGYL ?-D-MALTOSIDE 1 * 50 mg
Catalog # APOSBICL2509-50MG
Supplier: Apollo Scientific
Frequently Bought Together
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane, pure
Supplier: CHEMPUR
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Orthophosphoric acid 10%
Supplier: Hach
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
(±)-2-Ethyl-1-hexanol ≥99%
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.7% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99%
Supplier: Thermo Fisher Scientific