PROPARGYL ?-D-MALTOSIDE 1 * 25 mg
Catalog # APOSBICL2509-25MG
Supplier: Apollo Scientific
Frequently Bought Together
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Paraffin Oil heavy, liquid, Sigma-Aldrich®
Supplier: Merck
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
(±)-1,2-Propanediol, pure
Supplier: CHEMPUR
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Acetonitrile ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals