DECYL ACRYLATE 1 * 5 g
Catalog # APOSOR22270-5G
Supplier: Apollo Scientific
Frequently Bought Together
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethylene glycol, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran stabilized with 300 ppm of BHT for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Iodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific