WHEATON® COMPLETEPACK Sterile Components, Seals Only
Sterile aluminium hole seals with flip-off cap, 13 mm, Blue
Catalog # WHEAW988010-S
Supplier: DWK Life Sciences
WHEATON® COMPLETEPACK Sterile Components, Seals Only
Catalog # WHEAW988010-S
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Aluminium centre hole seals with polypropylene flip off caps, sterilised.
- Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
- Complete manufacturing and certificate of analysis traceability
- Ready-to-Use, Just-in-Time, minimum 180 day shelf life from time of shipment
- Container Closure Integrity (CCI) validation
USP <788>/EP 2.9.19 particulate matter in injections (vials and stoppers), USP <85>/EP 2.6.1, The Bacterial Endotoxins Test (vials and stoppers) and lot certified to SAL <10⁻⁶ (vials, stoppers, and seals), USP <660>/EP 3.2.1 glass containers for pharmaceutical use, and USP <381>/EP 3.2.9 elastomeric closures used in parenteral preparations.
Specifications
- Ø:13 mm
- Description:Sterile aluminium hole seals with flip-off cap
- Seq:02
- Cap colour:Blue