Order Entry
Poland
ContactUsLinkComponent
Flask, 100 ml, round bottom, center neck ST 24, angled side neck ST24/29 1 * 1 items
Catalog # WITG0.701.100.24
Supplier:  witeg Labortechnik
Flask, 100 ml, round bottom, center neck ST 24, angled side neck ST24/29 1 * 1 items
Catalog # WITG0.701.100.24
Supplier:  witeg Labortechnik
Frequently Bought Together