Order Entry
Poland
ContactUsLinkComponent
InfinityLab Poroshell 120 EC-C18, 4.6 x 100 mm, 4 um, analytical LC column, with column ID. Specifications:- Carbon Load: 10 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poros 1 * 1 items
Catalog # HEWL695970-902T
Dostawca:  Agilent
InfinityLab Poroshell 120 EC-C18, 4.6 x 100 mm, 4 um, analytical LC column, with column ID. Specifications:- Carbon Load: 10 %. Endc apped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 4.6 mm. LC Column Brand: InfinityLab Poros 1 * 1 items
Catalog # HEWL695970-902T
Dostawca:  Agilent
Frequently Bought Together