Order Entry
Poland
ContactUsLinkComponent
Specifications:- Compatible Column ID: 0.1-0.25 mm. Compatible Column OuterDiameter (OD): 0.36-0.39 mm. Ferrule Material: Stainless steel. Ferrule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.4 mm. Manufacturer Compatibility: A 1 * 10 items
Catalog # HEWLG2855-28501
Supplier:  Agilent
undefined
Specifications:- Compatible Column ID: 0.1-0.25 mm. Compatible Column OuterDiameter (OD): 0.36-0.39 mm. Ferrule Material: Stainless steel. Ferrule Type: Capillary. GC Component Type: Ferrule. Inner Diameter (ID): 0.4 mm. Manufacturer Compatibility: A 1 * 10 items
Catalog # HEWLG2855-28501
Supplier:  Agilent
Frequently Bought Together