Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Cole Parmer® SC-200 Sample Concentrators

Dri-Block® spacer inserts; 98,8×18,5×40,2 mm
  TECN36620-67
Cole Parmer® SC-200 Sample Concentrators
  TECN36620-67
Cole Parmer® SC-200 Sample Concentrators
Cole Parmer® SC-200 Sample Concentrators

Dri-Block® spacer inserts; 98,8×18,5×40,2 mm

 
166 zł
   
1 * 2 SZT
 

Quickly concentrate multiple samples in a block at once.

  • Variable needle position suits various Dri-Block® inserts
  • Small size makes it ideal for use under a fume hood
  • Needles, block, and block heater sold separately

Concentrator blows an inert gas over the surface of your samples via stainless steel needles (not included). Blown gas in combination with the heat from the heater produce ideal conditions for fast efficient evaporation. Insert needles into a silicone membrane in any configuration. Fully adjustable gas reservoir provides accurate height control. Maximum vertical traveling distance is 32 cm. Maximum gas usage is 15 L/min.

Informacje do zamówienia: Needles, block, and block heater sold separately. Order PTFE-coated needles for use with corrosive samples.

Informacja o dostawie: Supplied with gas reservoir and stand.

 
  • Description:
    Dri-Block® spacer inserts; 98,8×18,5×40,2 mm
  • Seq:
    06
Accessories

 

  •  
    Description
     
     
  • TECN36620-96
    Cole-Parmer sample concentrator needles, stainless steel, 76 mm; 100/pack
       
    1 * 100 SZT - 1 940 zł