Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
AMMONIUM MOLBYDATE TETRAHYDRATE CRYSTAL 1 * 25 g
Catalog # SPCMA1210-25GM
Supplier:
Spectrum Chemical
Expand image
AMMONIUM MOLBYDATE TETRAHYDRATE CRYSTAL 1 * 25 g
Catalog # SPCMA1210-25GM
Supplier:
Spectrum Chemical
Frequently Bought Together
Trifluoromethanesulphonic acid for synthesis
Supplier: Merck
Sodium L(+)-ascorbate, Sigma-Aldrich®
Supplier: Merck
Sodium hydrogen carbonate 99.5% for analysis
Supplier: Thermo Fisher Scientific
POTASU CHLORAN CZDA 1 * 500 g
Supplier: CHEMPUR
tri-Sodium citrate dihydrate, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Potassium cyanide 97% ACS
Supplier: Thermo Fisher Scientific
Mercury(II) sulphate ACS
Supplier: Thermo Fisher Scientific
Mercury(II) iodide, pure analytical reagent
Supplier: CHEMPUR
Sodium dodecyl sulphate (SDS), Sigma-Aldrich®
Supplier: Merck