BIORELEVANT FEDGAS PH6 BUFFER CONCENTRATE 6L 1 * 1 Bottle
Catalog # MSPPFDG-BUF6
Supplier: MARKET SOURCE PART PROCESS
BIORELEVANT FEDGAS PH6 BUFFER CONCENTRATE 6L 1 * 1 Bottle
Catalog # MSPPFDG-BUF6
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
BIORELEVANT FEDGAS GEL (1KG) MAKES 6L OF FED STATE GASTRIC DISSOLUTION MEDIA 1 * 1 Bottle
Supplier: MARKET SOURCE PART PROCESS
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
n-Heptane, pure
Supplier: CHEMPUR
n-Hexane for HPLC
Supplier: POCH AVANTOR BRAND
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific