J.T.Baker® Cell Lysis Solution, Biotech Reagent
Product Details & Documents
J.T.Baker® Cell Lysis Solution Biotech Reagent is a cGMP pre-formulated and ready to use detergent optimized for the extraction and recovery of AAV particles from producer cell lines (HEK293 parental and suspension lines, SF9). J.T.Baker® Cell Lysis Solution is a proprietary blend of nonionic surfactants, free of octyl phenol ethoxylate, which demonstrates comparable performance industry standard detergents while addressing environmental concerns associated with REACH and downstream clearance concerns. Designed for Gene Therapy workflows, this formulation is easy to use and handle as it is ready-to-use, has low viscosity, does not require cold temperature shipping or storage, and is available in sizes to support processes from development to large scale commercial batches. Detail description provided in technical literature.
- Comparable cell lysis performance to octyl phenol ethoxylate
- Improved protection of viral particles
- Easy to use and handle due to reduced foaming and viscosity and Room Termperature shipping and storage
- 0.1 µm filtered solution
Proprietary formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)
Chemical Specifications of GMP Cell lysis solution:
GMP Manufactured Product
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH : For information only
Viscosity (25°C) : For information only
Density: 0.9 – 1.2 g/mL
Total Aerobic Microbial Count: 100 cfu/mL
Total Yeast and Mold Count: 10 cfu/mL
UV-Vis (max Absorption) at 380-450 nm: For information only
Chemical Specifications of GMP Cell lysis solution
GMP Manufactured Product
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH: For information only
Viscosity (25°C): For information only
Density: 0.9 – 1.2 g/mL
Total Aerobic Microbial Count: 100 cfu/mL
Total Yeast and Mold Count: 10 cfu/mL
UV-Vis (max Absorption) at 380-450 nm: For information only
Documents
Certificates
Search by Lot NumberJ.T. Baker Cell Lysis Solution Detergent clearance using Affinity Chromatography J.T. Baker Cell Lysis Solution Detergent clearance using Anion Exchange Chromatography J.T.Baker® Cell Lysis Solution Efficiency J.T.Baker® Cell Lysis Solution for AAV Particle Protection J.T.Baker® Cell Lysis Solution for AAV Particle Protection, Biotech Reagent
Application Notes
J.T.Baker® Cell Lysis Solution, Biotech Reagent J.T.Baker® Cell Lysis Solution, Biotech Reagent J.T.Baker® Cell Lysis Solution, Biotech Reagent PDE Technical Note J.T.Baker® Cell Lysis Solution, Biotech Reagent Product Regulatory Datasheet J.T.Baker® Cell Lysis Solution, PDE Technical Note
Additional Product Details
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Description:Cell Lysis Solution, Biotech Reagent GMP
- Cat. No.:G193-05
- Environmental certification:EPA safer choice chemical
- Environmentally Preferable:Safer Human and Environmental Health
- Density:0.9-1.2 g/ml
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



