Clomifene for system suitability CRS 1 * 25 mg
Catalog # EDQMY0002178
Supplier: EDQM
Frequently Bought Together
UV/Visible standards, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Accessories for pH/°C meter, handheld
Supplier: TESTO
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Supplier: HAMILTON BONADUZ
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Supelco® COD CRM, standard solutions
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions in sachets
Supplier: Mettler - Toledo
Pararosaniline solution Reag. Ph. Eur. 1062201, decolourised
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach