0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
GLINU POTASU SIARCZAN 12 HYDRAT CZDA 1 * 25 g
CHMP114526603.0025
:
CHEMPUR
GLINU POTASU SIARCZAN 12 HYDRAT CZDA 1 * 25 g
CHMP114526603.0025
:
CHEMPUR
Frequently Bought Together
Aluminium ammonium sulphate dodecahydrate 99%, pure
: Thermo Fisher Scientific
Calcium carbonate 98+%, heavy powder, pure
: Thermo Fisher Scientific
Potassium bromide, pure analytical reagent
: CHEMPUR
Europium(III) chloride, anhydrous 99,9%
: Apollo Scientific
Ammonium heptamolybdate tetrahydrate, Sigma-Aldrich®
: Merck
Acetic acid glacial 99-100%, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
Ammonium hydrogen carbonate 98%, pure
: Thermo Fisher Scientific