3-ENOL ETHER CORTISONE ACETATE 1 * 100 mg
Catalog # MSPPSZH008065100MG
Supplier: MARKET SOURCE PART PROCESS
3-ENOL ETHER CORTISONE ACETATE 1 * 100 mg
Catalog # MSPPSZH008065100MG
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
2-Propanol, EMPROVE® EXPERT Ph. Eur., BP, USP for preparative chromatography, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Phenol liquified ≥90% for synthesis
Supplier: Merck
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Carbon disulphide ≥99.9%, Reagent Grade low in benzene
Supplier: Honeywell Chemicals
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
4-(Methylamino)phenol sulphate, pure analytical reagent
Supplier: POCH AVANTOR BRAND
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerine, Sigma-Aldrich®
Supplier: Merck