SODIUM BICARBONATE PURUM 1 * 25 kg
Catalog # CHMP428105306.9025
Supplier: CHEMPUR
Frequently Bought Together
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Disodium hydrogen phosphate dodecahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium hydrogen carbonate 99.7-100.3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals