Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MAG-BIND® PARTICLES HDQ - 200 ML 1 * 200 mL
Catalog # OMEGMBPHDQ-200
Supplier:
OMEGA BIO-TEK
Expand image
MAG-BIND® PARTICLES HDQ - 200 ML 1 * 200 mL
Catalog # OMEGMBPHDQ-200
Supplier:
OMEGA BIO-TEK
Frequently Bought Together
dNTP solutions, illustra™
Supplier: Cytiva
Recombinant protein of human thiosulfate sulfurtransferase (rhodanese) (TST), nuclear gene encoding mitochondrial protein 1 * 20 µG
Supplier: OriGene
Ready DNA Ultra 2.0 prefilled plates, for KingFisher Flex instruments, KingFisher™
Supplier: Thermo Fisher Scientific