CYNKU TLENEK CZDA 99,9% 1 * 1 kg
Catalog # CHMP112664701.1000
Supplier: CHEMPUR
Frequently Bought Together
Potassium acetate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sand (sea sand), extra pure, Supelco®
Supplier: Merck
Ammonium chloride 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium carbonate ≥97%, TECHNICAL
Supplier: VWR Chemicals
Iron(III) chloride 41% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Sodium hydrogen carbonate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium carbonate ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Potassium hydrogen phthalate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium carbonate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Zinc sulphate heptahydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
Supplier: VWR Chemicals