SODU FLUOREK R-R 0.1M 1L 1 * 1 L
CHMP817991998.1000
: CHEMPUR
Frequently Bought Together
Cell strainers for 1000 µl pipette tips, Flowmi™, SP Bel-Art
: Bel-Art Products, a Part of SP
Eppendorf® Assay/Reader Microplates
: EPPENDORF
Bioreactors, Wheaton® CELLine™
: DWK Life Sciences
tert-Butanol ≥99%
: Thermo Fisher Scientific
Cryoboxes B13/B15/B25
: GLW
ETHANOL 99,8% SOLUTION 1 * 100 mL
: CHEMPUR
VWR® Safe-T-Flex, Caps for Test Tubes
: VWR Collection
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
: Avantor Fluid Handling