Order Entry
Poland
ContactUsLinkComponent
Recombinant Human IL37 protein 1x 100 µg Purity: >= 95% SDS-PAGE Endotoxin level: <= 0.005 Eu/µg Suitable for: SDS-PAGE Protein length: Full length protein Animal free: Yes Carrier free: Yes Nature: Recombinant Species: Human Actual molecular weight: 1 * 100 µG
Catalog # ABCAAB287926-100
Supplier:  Abcam
Recombinant Human IL37 protein 1x 100 µg Purity: >= 95% SDS-PAGE Endotoxin level: <= 0.005 Eu/µg Suitable for: SDS-PAGE Protein length: Full length protein Animal free: Yes Carrier free: Yes Nature: Recombinant Species: Human Actual molecular weight: 1 * 100 µG
Catalog # ABCAAB287926-100
Supplier:  Abcam
Frequently Bought Together