Order Entry
Poland
Orders LinkContactUsLinkComponent
TIG Welding Gauntlets | Avantor
TIG Welding Gauntlets
XXL, Brown
numer katalogowy PRTWA521BRRXXL
Dostawca:   Portwest
TIG Welding Gauntlets
numer katalogowy PRTWA521BRRXXL
Dostawca:  Portwest
TIG Welding Gauntlets
TIG Welding Gauntlets
XXL, Brown
Przejdź do:
Price
66,60 zł
Zaloguj się dla dostępności
1 * 1 PARA
Product Details & Documents
The TIG Ultra Welding Gauntlet offers fantastic dexterity with a goat leather palm and excellent burn resistance due to the split leather backing. Designed for the professional TIG welder with aramid stitching for ultra durability.
  • EN12477 Type A welding protection
  • Premium quality leather welding gauntlet
  • The ultimate in dexterity for welders
  • Made from high quality durable goat leather
Ideal for welding and metal handling, these gloves feature reinforced Aramid stitching for maximum durability and burn protection.
Colour: Brown
Certifications: EN ISO 21420: 2020 Dexterity 2, EN 388: 2016 + A1: 2018 3243X, EN 12477: 2001 Type A, EN 407: 2020 422X31. ANSI/ISEA 105: 2016 CUT Level A2. CE certified.
Documents
Dane techniczne
  • Glove size:
    XXL
  • Colour:
    Brown
  • Cat. No.:
    PRTWA521BRRXXL
  • Latex / latex-free:
    Latex-free
  • Disposable / reusable:
    Reusable
  • Material:
    Leather/polyester
  • Powdered / powder-free:
    Powder-free
  • Sterility:
    Non sterile
  • Ambidextrous / hand-specific:
    Hand-specific